missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Orcein, pure

Catalog No. AC416571000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC416571000 100 g
AC416570050 5 g
AC416570250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC416571000 Supplier Thermo Scientific Chemicals Supplier No. 416571000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

orcein, C28H24N2O7, CAS Number-1400-62-0, Natural Red 28, 25g, 0.98 to 1.18, 06, 885, Black-Purple to Brown, 500.51, Pure, GHS Signal Word: Warning, GHS H Statement: Harmful if swallowed. Causes serious eye irritation. Causes skin irritation. | CAS: 1400-62-0 | C28H24N2O7 | 500.507 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1400-62-0
Molecular Formula C28H24N2O7
Molecular Weight (g/mol) 500.507
MDL Number MFCD00062310
InChI Key VPEASJIRGSVXBF-UHFFFAOYSA-N
Synonym Natural Red 28
PubChem CID 5386447
IUPAC Name 2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione
SMILES CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O

Specifications

Absorption Ratio 0.98 to 1.18
Infrared Spectrum Authentic
Lambda Max 575 to 582nm
Packaging Glass bottle
Beilstein 06, 885
Merck Index 15, 6958
Color Black/Purple to Brown
Quantity 100 g
Physical Form Powder
Formula Weight 500.51
Grade Pure
Chemical Name or Material orcein
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only