missing translation for 'onlineSavingsMsg'
Learn More

Orcein (Synthetic), TCI America™

Catalog No. O00611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O00611G Supplier TCI America Supplier No. O00611G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1400-62-0
Molecular Formula C28H24N2O7
Molecular Weight (g/mol) 500.507
MDL Number MFCD00062310
InChI Key VPEASJIRGSVXBF-UHFFFAOYSA-N
Synonym orcein, pacein, 2,8-bis 2,4-dihydroxy-6-methyl-anilino-1,9-dimethyl-dibenzofuran-3,7-dione, 2,8-bis 2,4-dihydroxy-6-methylanilino-1,9-dimethyldibenzo b,d furan-3,7-dione, unii-655waw52f4, orcein 1g, c.i. natural red 28, ncistruc1_000110, ncistruc2_001941, 2,4-dihydroxy-6-methylphenylamino-1,9-dimethyldibenzo b,d furan-3,7-dione
PubChem CID 5386447
IUPAC Name 2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione
SMILES CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O

Specifications

Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Orcein (Synthetic)