CAS RN 140-90-9
CAS RN 140-90-9
IUPAC Name:
sodium (ethoxymethanethioyl)sulfanide
Synonyms:
sodium ethyl xanthate
Molecular Weight (g/mol):
144.18
Molecular Formula:
C3H5NaOS2
InChi Key:
RZFBEFUNINJXRQ-UHFFFAOYSA-M
SMILES:
[Na+].CCOC([S-])=S
1
–
1
of
1
results
1
Sodium Ethylxanthate 97.0+%, TCI America™
CAS: 140-90-9 Molecular Formula: C3H5NaOS2 Molecular Weight (g/mol): 144.182 MDL Number: MFCD00068314 InChI Key: RZFBEFUNINJXRQ-UHFFFAOYSA-M Synonym: Ethylxanthic Acid Sodium Salt, Sodium O-Ethyl Dithiocarbonate, Sodium Xanthogenate PubChem CID: 23690437 IUPAC Name: sodium;ethoxymethanedithioate SMILES: CCOC(=S)[S-].[Na+]