accessibility menu, dialog, popup

UserName

CAS RN 140-39-6

IUPAC Name: 4-methylphenyl acetate
Synonyms: humibid guaiphenezine P-tolylacetate aether glycerinoguaiacolicus guaifenesine guaifenesina glycerol α-guiacyl ether p-tolyacetate trecid glycerol a-guiacyl ether
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: CDJJKTLOZJAGIZ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results