CAS RN 13963-57-0
Aluminum 2,4-pentanedionate
CAS: 13963-57-0 Molecular Formula: C15H21AlO6 Molecular Weight (g/mol): 324.31 MDL Number: MFCD00000013 InChI Key: KILURZWTCGSYRE-LNTINUHCSA-K Synonym: Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III) IUPAC Name: aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate) SMILES: [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Aluminum 2,4-pentanedionate, 99.995+% (metals basis)
CAS: 13963-57-0 Molecular Formula: C15H21AlO6 Molecular Weight (g/mol): 324.31 MDL Number: MFCD00000013 InChI Key: KILURZWTCGSYRE-LNTINUHCSA-K Synonym: Aluminum acetylacetonate IUPAC Name: aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate) SMILES: [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Aluminum acetylacetonate, 97%
CAS: 13963-57-0 Molecular Formula: C15H21AlO6 Molecular Weight (g/mol): 324.31 MDL Number: MFCD00000013 InChI Key: KILURZWTCGSYRE-LNTINUHCSA-K Synonym: aluminum 2, 4-pentanedionate IUPAC Name: aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate) SMILES: [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Tris(2,4-pentanedionato)aluminum(III) 99.0+%, TCI America™
CAS: 13963-57-0 Molecular Formula: C15H21AlO6 Molecular Weight (g/mol): 324.31 MDL Number: MFCD00000013 InChI Key: KILURZWTCGSYRE-LNTINUHCSA-K Synonym: Acetylacetone Aluminum(III) Salt, Aluminum(III) Acetylacetonate IUPAC Name: aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate) SMILES: [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O