accessibility menu, dialog, popup

UserName

CAS RN 13894-21-8

IUPAC Name: 1-(ethynesulfonyl)-4-methylbenzene
Synonyms: 1-(ethynesulfonyl)-4-methylbenzene
Molecular Weight (g/mol): 180.22
Molecular Formula: C9H8O2S
InChi Key: FTHKWIMQNXVEHW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results