accessibility menu, dialog, popup

UserName

HOBOHN

CAS RN 138642-62-3

IUPAC Name: (2-cyanophenyl)boronic acid
Synonyms: 2-cyanophenylboronic acid
Molecular Weight (g/mol): 146.94
Molecular Formula: C7H6BNO2
InChi Key: NPLZNDDFVCGRAG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1C#N
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)