accessibility menu, dialog, popup

UserName

CAS RN 13845-18-6

IUPAC Name: sodium sulfamate
Synonyms: sodium sulfamate
Molecular Weight (g/mol): 119.07
Molecular Formula: H2NNaO3S
InChi Key: QDWYPRSFEZRKDK-UHFFFAOYSA-M
SMILES: [Na+].NS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results