accessibility menu, dialog, popup

UserName

CAS RN 13820-53-6

IUPAC Name: disodium tetrachloropalladiumbis(ylium)
Synonyms: sodium tetrachloropalladate
Molecular Weight (g/mol): 294.2
Molecular Formula: Cl4Na2Pd
InChi Key: ABKQFSYGIHQQLS-UHFFFAOYSA-J
SMILES: [Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)