accessibility menu, dialog, popup

UserName

CAS RN 138113-08-3

IUPAC Name: 2-(7-methoxynaphthalen-1-yl)acetonitrile
Synonyms: 2-(7-methoxynaphthalen-1-yl)acetonitrile
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: PYJMGUQHJINLLD-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=C(CC#N)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results