accessibility menu, dialog, popup

UserName

CAS RN 137993-41-0

IUPAC Name: 16-cyano-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium perchlorate
Synonyms: 16-cyano-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium perchlorate
Molecular Weight (g/mol): 495.96
Molecular Formula: C26H26ClN3O5
InChi Key: HACOCUMLBPNDIN-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.N#CC1=C2C=C3CCC[N+]4=C3C(CCC4)=C2OC2=C3CCCN4CCCC(C=C12)=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results