accessibility menu, dialog, popup

UserName

OHS

CAS RN 13781-67-4

IUPAC Name: 2-(thiophen-3-yl)ethan-1-ol
Synonyms: 2-(thiophen-3-yl)ethanol
Molecular Weight (g/mol): 128.19
Molecular Formula: C6H8OS
InChi Key: YYPNNBPPDFTQFX-UHFFFAOYSA-N
SMILES: OCCC1=CSC=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)