accessibility menu, dialog, popup

UserName

CAS RN 13765-24-7

IUPAC Name: samarium(3+) trifluoride
Synonyms: samarium(III) fluoride
Molecular Weight (g/mol): 207.36
Molecular Formula: F3Sm
InChi Key: OJIKOZJGHCVMDC-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Sm+3]
Grade 
  • (3)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)