accessibility menu, dialog, popup

UserName

CAS RN 1375465-91-0

HNNNOHNOH

CAS RN 1375465-91-0

IUPAC Name: N-hydroxy-2-[(1-phenylcyclopropyl)amino]pyrimidine-5-carboxamide
Synonyms: N-hydroxy-2-[(1-phenylcyclopropyl)amino]pyrimidine-5-carboxamide
Molecular Weight (g/mol): 270.29
Molecular Formula: C14H14N4O2
InChi Key: LIIWIMDSZVNYHY-UHFFFAOYSA-N
SMILES: ONC(=O)C1=CN=C(NC2(CC2)C2=CC=CC=C2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results