accessibility menu, dialog, popup

UserName

CAS RN 137438-23-4

IUPAC Name: (11S)-3-chloro-11-methyl-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Synonyms: (11S)-3-chloro-11-methyl-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Molecular Weight (g/mol): 238.73
Molecular Formula: C11H11ClN2S
InChi Key: ZSDWARZBRRSRLT-LURJTMIESA-N
SMILES: C[C@H]1CCC2=C(C1)SC1=NC=NC(Cl)=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results