accessibility menu, dialog, popup

UserName

CH3NSHO

CAS RN 137-00-8

IUPAC Name: 2-(4-methyl-1,3-thiazol-5-yl)ethan-1-ol
Synonyms: 4-methyl-5-thiazoleethanol MHT C6H9nos hemineurine 4-methyl-5-hydroxethylthiazole
Molecular Weight (g/mol): 143.2
Molecular Formula: C6H9NOS
InChi Key: BKAWJIRCKVUVED-UHFFFAOYSA-N
SMILES: CC1=C(CCO)SC=N1
Boiling Point 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)