missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-thiazoleethanol, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018662
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018662 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018662 Supplier Merck Emd Millipore Supplier No. 61352100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 137-00-8
Density 1.196 g/mL (at 25°C)
Flash Point 112°C - closed cup
Physical Form Neat
Linear Formula C6H9 NOS
MDL Number MFCD00005339
Boiling Point 135°C (7 mmHg)
Synonym 5-(2-Hydroxyethyl)-4-methylthiazole; Sulfurol
Type Analytical Standard
Quantity 100 mg
Formula Weight 143.21
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less