missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Hydroxyethyl)-4-methylthiazole 98.0+%, TCI America™

Catalog No. H052725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H052725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H052725G Supplier TCI America Supplier No. H052725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-00-8
Molecular Formula C6H9NOS
Molecular Weight (g/mol) 143.204
MDL Number MFCD00005339
InChI Key BKAWJIRCKVUVED-UHFFFAOYSA-N
Synonym 4-methyl-5-thiazoleethanol, sulfurol, 5-2-hydroxyethyl-4-methylthiazole, hemineurine, 2-4-methylthiazol-5-yl ethanol, 4-methyl-5-thiazolethanol, 5-thiazoleethanol, 4-methyl, 2-4-methyl-1,3-thiazol-5-yl ethanol, 4-methyl-5-hydroxyethylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethan-1-ol
PubChem CID 1136
ChEBI CHEBI:17957
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethanol
SMILES CC1=C(SC=N1)CCO

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
Formula Weight 143.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-(2-Hydroxyethyl)-4-methylthiazole