CAS RN 136831-48-6
IUPAC Name:
(2Z)-3-(3,4-dimethoxyphenyl)-2-(pyridin-3-yl)prop-2-enenitrile
Synonyms:
(2Z)-3-(3,4-dimethoxyphenyl)-2-(pyridin-3-yl)prop-2-enenitrile
Molecular Weight (g/mol):
266.3
Molecular Formula:
C16H14N2O2
InChi Key:
DBGZNJVTHYFQJI-RIYZIHGNSA-N
SMILES:
COC1=CC=C(\C=C(/C#N)C2=CC=CN=C2)C=C1OC
1
–
1
of
1
results
1
Tyrphostin RG 13022 98.0+%, TCI America™
CAS: 136831-48-6 Molecular Formula: C16H14N2O2 Molecular Weight (g/mol): 266.3 MDL Number: MFCD00236444 InChI Key: DBGZNJVTHYFQJI-RIYZIHGNSA-N Synonym: 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrile PubChem CID: 5468216 IUPAC Name: (Z)-3-(3,4-dimethoxyphenyl)-2-pyridin-3-ylprop-2-enenitrile SMILES: COC1=C(C=C(C=C1)C=C(C#N)C2=CN=CC=C2)OC