accessibility menu, dialog, popup

UserName

BOOH3CH3CH3CH3CSNOH3CCH3SBOOCH3CH3H3CCH3ONCH3CH3

CAS RN 1351986-34-9

IUPAC Name: 2,5-bis(2-octyldodecyl)-3,6-bis[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
Synonyms: 2,5-bis(2-octyldodecyl)-3,6-bis[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]pyrrolo[3,4-c]pyrrole-1,4-dione
Molecular Weight (g/mol): 1113.35
Molecular Formula: C66H110B2N2O6S2
InChi Key: OZQFATKGMJQJSF-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC(CCCCCCCC)CN1C(=O)C2=C(N(CC(CCCCCCCC)CCCCCCCCCC)C(=O)C2=C1C1=CC=C(S1)B1OC(C)(C)C(C)(C)O1)C1=CC=C(S1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results