accessibility menu, dialog, popup

UserName

CAS RN 135053-11-1

IUPAC Name: (1S,2R)-2-carbamoylcyclopentan-1-aminium
Synonyms: (1S,2R)-2-carbamoylcyclopentan-1-aminium
Molecular Weight (g/mol): 129.18
Molecular Formula: C6H13N2O
InChi Key: FUGFTUCRJJFPES-UHNVWZDZSA-O
SMILES: NC(=O)[C@@H]1CCC[C@@H]1[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)