CAS RN 135034-10-5
IUPAC Name:
3-chloro-6-iodopyridazine
Synonyms:
3-chloro-6-iodopyridazine
Molecular Weight (g/mol):
240.43
Molecular Formula:
C4H2ClIN2
InChi Key:
PNEPCDPKMXJYIQ-UHFFFAOYSA-N
SMILES:
ClC1=NN=C(I)C=C1
1
–
1
of
1
results
1
Promotions Available
3-Chloro-6-iodopyridazine, 95%
3-Chloro-6-iodopyridazine, 95%
CAS: 135034-10-5 Molecular Formula: C4H2ClIN2 Molecular Weight (g/mol): 240.43 MDL Number: MFCD08275187 InChI Key: PNEPCDPKMXJYIQ-UHFFFAOYSA-N Synonym: 3-chloro-6-iodo-pyridazine,pyridazine, 3-chloro-6-iodo,3-chloro-6-pyrazine,acmc-1cg9b,ksc169s6d,3-chloranyl-6-iodanyl-pyridazine,6-chloro-3-iodopyridazine,3-chloro-6-iodo-1,2-diazine PubChem CID: 15418839 IUPAC Name: 3-chloro-6-iodopyridazine SMILES: ClC1=NN=C(I)C=C1