accessibility menu, dialog, popup

UserName

CoCoCoOPOOOOPOOO

CAS RN 13455-36-2

IUPAC Name: tris(λ²-cobalt(2+)) diphosphate
Synonyms: cobalt phosphate
Molecular Weight (g/mol): 366.74
Molecular Formula: Co3O8P2
InChi Key: ZBDSFTZNNQNSQM-UHFFFAOYSA-H
SMILES: [Co++].[Co++].[Co++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results