accessibility menu, dialog, popup

UserName

CAS RN 13380-94-4

O

CAS RN 13380-94-4

IUPAC Name: tricyclo[5.2.1.0²,⁶]decan-8-one
Synonyms: tricyclodecane-8-one
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: OMIDXVJKZCPKEI-UHFFFAOYNA-N
SMILES: O=C1CC2CC1C1CCCC21
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results