accessibility menu, dialog, popup

UserName

CAS RN 133745-75-2

IUPAC Name: N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
Synonyms: N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
Molecular Weight (g/mol): 315.33
Molecular Formula: C12H10FNO4S2
InChi Key: RLKHFSNWQCZBDC-UHFFFAOYSA-N
SMILES: FN(S(=O)(=O)C1=CC=CC=C1)S(=O)(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (4)