missing translation for 'onlineSavingsMsg'
Learn More

N-Fluorobenzenesulfonimide, 97%

Catalog No. AC311900250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC311900250 25 g Glass bottle
AC311900050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC311900250 Supplier Thermo Scientific Chemicals Supplier No. 311900250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 133745-75-2 | C12H10FNO4S2 | 315.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 133745-75-2
Molecular Formula C12H10FNO4S2
Molecular Weight (g/mol) 315.35
InChI Key RLKHFSNWQCZBDC-UHFFFAOYSA-N
Synonym n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
PubChem CID 588007
IUPAC Name N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 110.0°C to 115.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Merck Index 15,4202
Formula Weight 315.35
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-Fluorobenzenesulfonimide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only