accessibility menu, dialog, popup

UserName

CAS RN 133073-73-1

IUPAC Name: (2S)-2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioic acid
Synonyms: methotrexate|L(+)-Amethopterin hydrate
Molecular Weight (g/mol): 454.45
Molecular Formula: C20H22N8O5
InChi Key: FBOZXECLQNJBKD-UGPWUYPHNA-N
SMILES: CN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)N[C@@H](CCC(O)=O)C(O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results