accessibility menu, dialog, popup

UserName

CAS RN 13294-71-8

IUPAC Name: (2E)-2-bromobut-2-ene
Synonyms: 2-bromo-2-butene (cis)
Molecular Weight (g/mol): 135
Molecular Formula: C4H7Br
InChi Key: UILZQFGKPHAAOU-ONEGZZNKSA-N
SMILES: C\C=C(/C)Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)