accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OH2OH2OYYOOOOOOOOOOOO

CAS RN 13266-82-5

IUPAC Name: diyttrium(3+) nonahydrate trioxalate
Synonyms: diyttrium(3+) nonahydrate trioxalate
Molecular Weight (g/mol): 604
Molecular Formula: C6H18O21Y2
InChi Key: CTXJKXXQOORSKU-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.O.[Y+3].[Y+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results