accessibility menu, dialog, popup

UserName

CAS RN 132-86-5

IUPAC Name: naphthalene-1,3-diol
Synonyms: naphthalene-1,3-diol
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: XOOMNEFVDUTJPP-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (5)