accessibility menu, dialog, popup

UserName

CAS RN 132-27-4

IUPAC Name: sodium [1,1'-biphenyl]-2-olate
Synonyms: sodium ortho-phenylphenate
Molecular Weight (g/mol): 192.19
Molecular Formula: C12H9NaO
InChi Key: KSQXVLVXUFHGJQ-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC=CC=C1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results