accessibility menu, dialog, popup

UserName

ClClNOOCl

CAS RN 13167-25-4

IUPAC Name: 1-(2,4,6-trichlorophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(2,4,6-trichlorophenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 276.5
Molecular Formula: C10H4Cl3NO2
InChi Key: VHZJMAJCUAWIHV-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(N2C(=O)C=CC2=O)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results