Learn More
Ytterbium(III) oxide, REacton™, 99.99% (REO)
Description
Ytterbium(III) oxide serves as a dopant for silicate mineral crystals in lasers, and a colorant for glasses and enamels. It is employed in the preparation of optical fibers, and special types of alloys. It is used in the preparation of chiral ytterbium complexes which are used as catalysts for enantioselective organic synthesis with high yields. It finds application in optical, electronic and ceramic industries. It is used as additives for fluorescent powder and in the preparation of magnetic bubbles in computers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2227°C |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.99% (REO) |
| Density | 9.2 g/mL |
| Merck Index | 14,10105 |
| Solubility Information | Soluble in hot dilute acids. Insoluble in water. |
| Formula Weight | 394.08 |
| Percent Purity | 99.99% |
| Grade | Reacton |
| Show More |
Safety and Handling
| EINECSNumber | 215-234-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only