missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) Oxide 98.0+%, TCI America™

Catalog No. Y00085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y00085G 5 g
Y000825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Y00085G Supplier TCI America Supplier No. Y00085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1314-37-0
Molecular Formula O3Yb2
Molecular Weight (g/mol) 394.09
MDL Number MFCD00011290
InChI Key UZLYXNNZYFBAQO-UHFFFAOYSA-N
IUPAC Name diytterbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Yb+3].[Yb+3]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 394.11
Percent Purity ≥98.0% (T)
Chemical Name or Material Ytterbium(III) Oxide
EINECSNumber (1)-0824
TSCA Yes