accessibility menu, dialog, popup

UserName

CAS RN 13130-79-5

IUPAC Name: 1-bromoisoquinolin-3-amine
Synonyms: 1-bromoisoquinolin-3-amine
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: PSQUIUNIVDKHJK-UHFFFAOYSA-N
SMILES: NC1=NC(Br)=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results