accessibility menu, dialog, popup

UserName

CAS RN 1313-59-3

ONaNa

CAS RN 1313-59-3

IUPAC Name: disodium oxidandiide
Synonyms: sodium oxide
Molecular Weight (g/mol): 61.98
Molecular Formula: Na2O
InChi Key: KKCBUQHMOMHUOY-UHFFFAOYSA-N
SMILES: [O--].[Na+].[Na+]
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results