missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxide

Catalog No. AA3671222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3671222 100 g
AA3671214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3671222 Supplier Thermo Scientific Chemicals Supplier No. 03671222
Only null left

CAS: 1313-59-3 | Na2O | 61.979 g/mol

Sodium oxide is used in chemical manufacturing, ceramics and glasses. Ethylbenzene dehydrogenation and carbon dioxide shift-reaction occurs using a sodium oxide/alumina catalyst. It is employed as catalyzing gasification of CO2 on carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-59-3
Molecular Formula Na2O
Molecular Weight (g/mol) 61.979
MDL Number MFCD00046201
InChI Key KKCBUQHMOMHUOY-UHFFFAOYSA-N
Synonym sodium oxide, disodium oxide, disodium monoxide, disodium oxygen-2 anion, disodium oxidandiide, sodium oxide 25g, disodium oxygen 2-, 2na.o
PubChem CID 73971
IUPAC Name disodium;oxygen(2-)
SMILES [O-2].[Na+].[Na+]

Specifications

Melting Point >400°C (decomposition)
Color White to Gray
Physical Form Beads
Quantity 100 g
UN Number UN1825
Merck Index 14,8651
Solubility Information Soluble in water.
Formula Weight 61.98
Odor Odorless
Sensitivity Air and moisture sensitive
Density 2.27 g/mL
Chemical Name or Material Sodium oxide
Show More Show Less
Hazard Category H271-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: I
EINECSNumber 215-208-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only