accessibility menu, dialog, popup

UserName

CAS RN 13114-90-4

IUPAC Name: (2-bromophenyl)urea
Synonyms: 2-bromophenylurea
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: QXEDXIJDCOADGG-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=CC=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results