accessibility menu, dialog, popup

UserName

CAS RN 131-91-9

IUPAC Name: 1-nitrosonaphthalen-2-ol
Synonyms: 1-nitroso-2-naphthol
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: YXAOOTNFFAQIPZ-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1N=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)