missing translation for 'onlineSavingsMsg'
Learn More

1-Nitroso-2-naphthol, 98%

Catalog No. AAA1062230
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062230 250 g
AAA1062209 10 g
AAA1062218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1062230 Supplier Thermo Scientific Chemicals Supplier No. A1062230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131-91-9 | C10H7NO2 | 173.171 g/mol

1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 131-91-9
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00003884
InChI Key YXAOOTNFFAQIPZ-UHFFFAOYSA-N
Synonym 1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
PubChem CID 8580
IUPAC Name 1-nitrosonaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2N=O)O

Specifications

Melting Point 104°C to 108°C (decomposition)
Quantity 250 g
UN Number UN3077
Beilstein 776947
Merck Index 14,6641
Solubility Information Insoluble in water.
Formula Weight 173.17
Percent Purity 99%
Chemical Name or Material 1-Nitroso-2-naphthol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 205-043-0
RTECSNumber QL4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only