missing translation for 'onlineSavingsMsg'
Learn More

1-Nitroso-2-naphthol 98.0+%, TCI America™

Catalog No. N026625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N026625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N026625G Supplier TCI America Supplier No. N026625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 131-91-9
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00003884
InChI Key YXAOOTNFFAQIPZ-UHFFFAOYSA-N
Synonym 1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
PubChem CID 8580
IUPAC Name 1-nitrosonaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2N=O)O

Specifications

Melting Point 107°C
Quantity 25 g
Formula Weight 173.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Nitroso-2-naphthol
EINECSNumber (4)-0370
RTECSNumber QL4725000
TSCA Yes