CAS RN 130000-40-7
IUPAC Name:
N-[2,6-dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxamide
Synonyms:
thifluzamide
Molecular Weight (g/mol):
528.06
Molecular Formula:
C13H6Br2F6N2O2S
InChi Key:
WOSNCVAPUOFXEH-UHFFFAOYSA-N
SMILES:
CC1=NC(=C(S1)C(=O)NC1=C(Br)C=C(OC(F)(F)F)C=C1Br)C(F)(F)F
1
–
1
of
1
results
1
Thifluzamide 98.0+%, TCI America™
CAS: 130000-40-7 Molecular Formula: C13H6Br2F6N2O2S Molecular Weight (g/mol): 528.061 MDL Number: MFCD03428071 InChI Key: WOSNCVAPUOFXEH-UHFFFAOYSA-N Synonym: N-[2,6-Dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)thiazole-5-carboxamide PubChem CID: 86389 ChEBI: CHEBI:81794 IUPAC Name: N-[2,6-dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxamide SMILES: CC1=NC(=C(S1)C(=O)NC2=C(C=C(C=C2Br)OC(F)(F)F)Br)C(F)(F)F