accessibility menu, dialog, popup

UserName

OHOHOOHOHO

CAS RN 130-85-8

IUPAC Name: 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
Synonyms: pamoic acid pamaquine naphthoate pamosaeure embonic acid
Molecular Weight (g/mol): 388.38
Molecular Formula: C23H16O6
InChi Key: WLJNZVDCPSBLRP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)