accessibility menu, dialog, popup

UserName

CAS RN 129283-82-5

CH3CH2OSSSOCH2H3C

CAS RN 129283-82-5

IUPAC Name: 2-methyl-1-{[4-({4-[(2-methylprop-2-enoyl)sulfanyl]phenyl}sulfanyl)phenyl]sulfanyl}prop-2-en-1-one
Synonyms: 2-methyl-1-{[4-({4-[(2-methylprop-2-enoyl)sulfanyl]phenyl}sulfanyl)phenyl]sulfanyl}prop-2-en-1-one
Molecular Weight (g/mol): 386.54
Molecular Formula: C20H18O2S3
InChi Key: SPNAQSNLZHHUIJ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)SC1=CC=C(SC2=CC=C(SC(=O)C(C)=C)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results