accessibility menu, dialog, popup

UserName

NBr

CAS RN 129013-83-8

IUPAC Name: 3-(4-bromophenyl)pyridine
Synonyms: 3-(4-bromophenyl)pyridine
Molecular Weight (g/mol): 234.1
Molecular Formula: C11H8BrN
InChi Key: FCHUOBPHXDXZBK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CN=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results