accessibility menu, dialog, popup

Test

CAS RN 128607-22-7

CAS RN 128607-22-7

IUPAC Name: 2-{4-[(1Z)-4-chloro-1,2-diphenylbut-1-en-1-yl]phenoxy}ethan-1-ol
Synonyms: ospemifene
Molecular Weight (g/mol): 378.9
Molecular Formula: C24H23ClO2
InChi Key: LUMKNAVTFCDUIE-VHXPQNKSSA-N
SMILES: OCCOC1=CC=C(C=C1)C(=C(\CCCl)C1=CC=CC=C1)\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results