accessibility menu, dialog, popup

UserName

FBrN

CAS RN 128071-98-7

IUPAC Name: 4-bromo-2-fluoropyridine
Synonyms: 4-bromo-2-fluoropyridine
Molecular Weight (g/mol): 175.99
Molecular Formula: C5H3BrFN
InChi Key: PTPTZLXZHPPVKG-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results