accessibility menu, dialog, popup

UserName

CAS RN 128055-74-3

BrBrBrBr

CAS RN 128055-74-3

IUPAC Name: 2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene]
Synonyms: 2,2',7,7'-tetrabromo-9,9'-spirobi[fluorene]
Molecular Weight (g/mol): 631.99
Molecular Formula: C25H12Br4
InChi Key: MASXXNUEJVMYML-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C21C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results