accessibility menu, dialog, popup

UserName

CAS RN 127946-77-4

IUPAC Name: 1-aminocyclopropane-1-carbonitrile hydrochloride
Synonyms: 1-aminocyclopropane-1-carbonitrile hydrochloride
Molecular Weight (g/mol): 118.56
Molecular Formula: C4H7ClN2
InChi Key: PCEIEQLJYDMRFZ-UHFFFAOYSA-N
SMILES: Cl.NC1(CC1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)