accessibility menu, dialog, popup

UserName

ON

CAS RN 127406-56-8

IUPAC Name: 4-(pyridin-2-yl)benzaldehyde
Synonyms: 4-(pyridin-2-yl)benzaldehyde
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: NMLYGLCBSFKJFI-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)